Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(3,5-Dichlorophenyl)-2,5-dimethylpyrrole, 97%

Click to view available options
Quantity:
1g
5g
25g
175205-50-2
C12H11Cl2N
240.127
MFCD00084999
PZQGRGDARNXLSP-UHFFFAOYSA-N
1-3,5-dichlorophenyl-2,5-dimethyl-1h-pyrrole,1-3,5-dichlorophenyl-2,5-dimethylpyrrole,acmc-1ccua,1h-pyrrole,1-3,5-dichlorophenyl-2,5-dimethyl
10490192
1-(3,5-dichlorophenyl)-2,5-dimethylpyrrole
CC1=CC=C(N1C2=CC(=CC(=C2)Cl)Cl)C
175205-50-2
C12H11Cl2N
240.127
MFCD00084999
PZQGRGDARNXLSP-UHFFFAOYSA-N
1-3,5-dichlorophenyl-2,5-dimethyl-1h-pyrrole,1-3,5-dichlorophenyl-2,5-dimethylpyrrole,acmc-1ccua,1h-pyrrole,1-3,5-dichlorophenyl-2,5-dimethyl
10490192
1-(3,5-dichlorophenyl)-2,5-dimethylpyrrole
CC1=CC=C(N1C2=CC(=CC(=C2)Cl)Cl)C
Quantity 25g
Melting Point 78°C to 80°C
Formula Weight 240.13
Percent Purity 97%
Chemical Name or Material 1-(3,5-Dichlorophenyl)-2,5-dimethylpyrrole