Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,5-Dichlorophenyl)-2-hydroxyethylamine, 94%, Acros Organics

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
372144-00-8
C8H9Cl2NO
206.07
LRORZXIRKBZZAG-UHFFFAOYSA-N
2-amino-2-3,5-dichlorophenyl ethanol,1-3,5-dichlorophenyl-2-hydroxyethylamine,2-amino-2-3,5-dichlorophenyl ethan-1-ol,benzeneethanol, b-amino-3,5-dichloro
2734585
2-amino-2-(3,5-dichlorophenyl)ethanol
C1=C(C=C(C=C1Cl)Cl)C(CO)N
372144-00-8
C8H9Cl2NO
206.07
LRORZXIRKBZZAG-UHFFFAOYSA-N
2-amino-2-3,5-dichlorophenyl ethanol,1-3,5-dichlorophenyl-2-hydroxyethylamine,2-amino-2-3,5-dichlorophenyl ethan-1-ol,benzeneethanol, b-amino-3,5-dichloro
2734585
2-amino-2-(3,5-dichlorophenyl)ethanol
C1=C(C=C(C=C1Cl)Cl)C(CO)N
Quantity 500mg
Packaging Glass bottle
Assay Percent Range 93% min. (HPLC)
Formula Weight 206.07
Percent Purity 94%
Chemical Name or Material 1-(3, 5-Dichlorophenyl)-2-hydroxyethylamine

GHS Signal Word: Warning