Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3,5-Triaza-7-phosphaadamantane, ≥97%

Click to view available options
Quantity:
1g
5g
53597-69-6
C6H12N3P
157.157
MFCD00154905
FXXRPTKTLVHPAR-UHFFFAOYSA-N
1,3,5-triaza-7-phosphaadamantane,1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane,1,3,5-triaza-7-phosphaadamantane,min,1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane,acmc-20ap5q,1,5-triaza-9-phospha-adamantane,1,3,5-triaza-7-phosphaadamantane,1,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane,1,3,5-triaza-7-phosphatricyclo 3.3.1.13.7 decane,1,3,5-triaza-7-phosphatricyclo 3.3.1.1 3,7 decane
143061
C1N2CN3CN1CP(C2)C3
53597-69-6
C6H12N3P
157.157
MFCD00154905
FXXRPTKTLVHPAR-UHFFFAOYSA-N
1,3,5-triaza-7-phosphaadamantane,1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane,1,3,5-triaza-7-phosphaadamantane,min,1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane,acmc-20ap5q,1,5-triaza-9-phospha-adamantane,1,3,5-triaza-7-phosphaadamantane,1,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane,1,3,5-triaza-7-phosphatricyclo 3.3.1.13.7 decane,1,3,5-triaza-7-phosphatricyclo 3.3.1.1 3,7 decane
143061
C1N2CN3CN1CP(C2)C3
Quantity 1g
Color White
Sensitivity Hygroscopic
Melting Point 244°C to 250°C
Formula Weight 157.15
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material 1,3,5-Triaza-7-phosphaadamantane