Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tribromobenzene, 98%, Thermo Scientific Chemicals

Catalog Number AAB2386014
Click to view available options
Quantity:
25 g
100 g
500 g
626-39-1
C6H3Br3
314.80
MFCD00000080
YWDUZLFWHVQCHY-UHFFFAOYSA-N
benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o
12279
1,3,5-tribromobenzene
BrC1=CC(Br)=CC(Br)=C1
626-39-1
C6H3Br3
314.80
MFCD00000080
YWDUZLFWHVQCHY-UHFFFAOYSA-N
benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o
12279
1,3,5-tribromobenzene
BrC1=CC(Br)=CC(Br)=C1

1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

Solubility
Soluble in hot ethanol and acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 121°C to 124°C
Boiling Point 271°C
Quantity 25 g
Beilstein 1858917
Solubility Information Soluble in hot ethanol and acetic acid. Insoluble in water.
Formula Weight 314.8
Percent Purity 98%
Chemical Name or Material 1,3,5-Tribromobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-947-3
RTECSNumber DC1957200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit