Learn More
1,3,5-Tribromobenzene, 98%, Thermo Scientific Chemicals
Description
1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.
Solubility
Soluble in hot ethanol and acetic acid. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 121°C to 124°C |
Boiling Point | 271°C |
Quantity | 25 g |
Beilstein | 1858917 |
Solubility Information | Soluble in hot ethanol and acetic acid. Insoluble in water. |
Formula Weight | 314.8 |
Percent Purity | 98% |
Chemical Name or Material | 1,3,5-Tribromobenzene |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 210-947-3 |
RTECSNumber | DC1957200 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.