Alfa Aesar™ 1,3,5-Trichloro-2-nitrobenzene, 98+%

Manufacturer: Alfa Aesar™ A1226222

Quantity 100g

 View more versions of this product

Catalog No. AAA1226222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18708-70-8
    Molecular Formula C6H2Cl3NO2
    Formula Weight 226.45
    MDL Number MFCD00014690
    UN Number UN2811
    Beilstein 2213282
    Synonym 2-Nitro-1,3,5-trichlorobenzene; 2,4,6-Trichloronitrobenzene
    Chemical Name or Material 1,3,5-Trichloro-2-nitrobenzene

    Specifications

    Formula Weight 226.45
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 2213282
    Synonym 2-Nitro-1,3,5-trichlorobenzene; 2,4,6-Trichloronitrobenzene
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Molecular Formula C6H2Cl3NO2
    MDL Number MFCD00014690
    CAS 18708-70-8
    UN Number UN2811
    Health Hazard 1 Danger
    Melting Point 68° to 73°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 100g
    Chemical Name or Material 1,3,5-Trichloro-2-nitrobenzene
    Assay ≥98%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit