Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3,5-Triethynylbenzene, 98%

Click to view available options
Quantity:
1g
5g
7567-63-7
C12H6
150.18
MFCD00068621
ZDRMMTYSQSIGRY-UHFFFAOYSA-N
benzene, 1,3,5-triethynyl,benzene,1,3,5-triethynyl,1,3,5-phenylene-triacetylene,acmc-209ozh,1,3,5-triethynyl-benzene,1,3,5-triethynylbenzene,1,3,5-triethynylbenzene 1g,benzene, 1,3,5-triethynyl-8ci, 9ci
139048
1,3,5-triethynylbenzene
C#CC1=CC(=CC(=C1)C#C)C#C
7567-63-7
C12H6
150.18
MFCD00068621
ZDRMMTYSQSIGRY-UHFFFAOYSA-N
benzene, 1,3,5-triethynyl,benzene,1,3,5-triethynyl,1,3,5-phenylene-triacetylene,acmc-209ozh,1,3,5-triethynyl-benzene,1,3,5-triethynylbenzene,1,3,5-triethynylbenzene 1g,benzene, 1,3,5-triethynyl-8ci, 9ci
139048
1,3,5-triethynylbenzene
C#CC1=CC(=CC(=C1)C#C)C#C
Applications
1,3,5-Triethynylbenzene is used as the carbon precursor for graphene synthesis on rhodium. It is also used as a connecting unit for various transition metal building blocks.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
Melting Point 105°C to 107°C
Quantity 1g
UN Number UN1325
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 1,3,5-Triethynylbenzene

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.