Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3,5-Trifluoro-2-nitrobenzene, 98%

Click to view available options
Quantity:
1g
5g
25g
315-14-0
C6H2F3NO2
177.082
MFCD00014687
PWRFDGYYJWQIAB-UHFFFAOYSA-N
2,4,6-trifluoronitrobenzene,2,2,6-trifluoronitrobenzene,benzene, 1,3,5-trifluoro-2-nitro,1,3,5-trifluoro-2-nitro-benzene,2-nitro-1,3,5-trifluorobenzene,pubchem10646,acmc-209hn4,1,3,5-trifluoronitrobenzene,2,4,6 trifluoronitrobenzene,ksc226o4t
67567
1,3,5-trifluoro-2-nitrobenzene
C1=C(C=C(C(=C1F)[N+](=O)[O-])F)F
315-14-0
C6H2F3NO2
177.082
MFCD00014687
PWRFDGYYJWQIAB-UHFFFAOYSA-N
2,4,6-trifluoronitrobenzene,2,2,6-trifluoronitrobenzene,benzene, 1,3,5-trifluoro-2-nitro,1,3,5-trifluoro-2-nitro-benzene,2-nitro-1,3,5-trifluorobenzene,pubchem10646,acmc-209hn4,1,3,5-trifluoronitrobenzene,2,4,6 trifluoronitrobenzene,ksc226o4t
67567
1,3,5-trifluoro-2-nitrobenzene
C1=C(C=C(C(=C1F)[N+](=O)[O-])F)F
Flash Point 77°C (170°F)
Quantity 1g
Density 1.514
Refractive Index 1.479
Melting Point 2°C to 5°C
Boiling Point 171°C to 172°C
UN Number UN2810
Beilstein 2106869
Formula Weight 177.08
Percent Purity 98%
Chemical Name or Material 1,3,5-Trifluoro-2-nitrobenzene
Show More Show Less

GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 206-248-8

TSCA : No

Recommended Storage : Ambient temperatures