Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Triisopropylbenzene, 95%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
717-74-8
C15H24
204.35
MFCD00008890
VUMCUSHVMYIRMB-UHFFFAOYSA-N
1,3,5-triisopropylbenzene,2,4,6-triisopropylbenzene,benzene, 1,3,5-tris 1-methylethyl,1,3,5-tris propan-2-yl benzene,unii-fr9y346wpb,benzene, 1,3,5-triisopropyl,fr9y346wpb,benzene, tris 1-methylethyl,pubchem13720,1,5-triisopropylbenzene
12860
1,3,5-tri(propan-2-yl)benzene
CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C
717-74-8
C15H24
204.35
MFCD00008890
VUMCUSHVMYIRMB-UHFFFAOYSA-N
1,3,5-triisopropylbenzene,2,4,6-triisopropylbenzene,benzene, 1,3,5-tris 1-methylethyl,1,3,5-tris propan-2-yl benzene,unii-fr9y346wpb,benzene, 1,3,5-triisopropyl,fr9y346wpb,benzene, tris 1-methylethyl,pubchem13720,1,5-triisopropylbenzene
12860
1,3,5-tri(propan-2-yl)benzene
CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C
Specific Gravity 0.84
Flash Point 86°C
Boiling Point 232°C to 236°C
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4874 to 1.4894
Density 0.8400g/mL
Color Colorless
Quantity 100mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula C6H3[CH(CH3)2]3
Beilstein 05, 458
Formula Weight 204.35
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 1, 3, 5-Triisopropylbenzene, 97%
Show More Show Less

EINECSNumber : 211-941-3

TSCA : TSCA