Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3,5-Trimethoxy-2-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ A1047306

Quantity 5g

 View more versions of this product

Catalog No. AAA1047306


  Check Availability
Add to cart

Specifications

Description

Applications
2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

Solubility
Soluble in water.

Notes
Desiccate at room temperature. Incompatabile to strong oxidizing agents, strong acids and bases. Store in a cool, dry, well-ventilated place.
Description & Specifications

Chemical Identifiers

14227-18-0
213.19
2277262
1,3,5-Trimethoxy-2-nitrobenzene
C9H11NO5
MFCD00016992
2-Nitro-1,3,5-trimethoxybenzene; 2,4,6-Trimethoxynitrobenzene

Specifications

150° to 153°C
14227-18-0
213.19
2277262
1,3,5-Trimethoxy-2-nitrobenzene
5g
C9H11NO5
MFCD00016992
2-Nitro-1,3,5-trimethoxybenzene; 2,4,6-Trimethoxynitrobenzene
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit