Alfa Aesar™ 1,3,5-Trimethyl-1,4-cyclohexadiene, 98%

Manufacturer: Alfa Aesar™ H2592106

Quantity 5g

 View more versions of this product

Catalog No. AAH2592106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4074-23-1
    Molecular Formula C9H14
    Formula Weight 122.21
    MDL Number MFCD04974021
    UN Number UN3295
    Chemical Name or Material 1,3,5-Trimethyl-1,4-cyclohexadiene

    Specifications

    Formula Weight 122.21
    UN Number UN3295
    Molecular Formula C9H14
    MDL Number MFCD04974021
    Health Hazard 1 Warning
    Flash Point 35°C (95°F)
    Boiling Point 172°C
    Quantity 5g
    CAS 4074-23-1
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Chemical Name or Material 1,3,5-Trimethyl-1,4-cyclohexadiene
    Assay 98%

    Description

    Applications
    1,3,5-Trimethyl-1,4-cyclohexadiene is used as intermediates in pharmaceutical industry.

    Solubility
    Insoluble in water.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit