Learn More
Medchemexpress LLC 1,3,5-trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene | 1709-70-2 | 100.0% | 25 G
SDP
Supplier:  Medchemexpress LLC HY12038825G
Encompass_Preferred
Irganox 1330 is a phenolic antioxidant (1,3,5-trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene) used to protect polymeric materials from thermo-oxidative degradation and discoloration. It is supplied as a solid and is commonly incorporated at low concentrations into polyolefins and other plastics to improve processing stability and resistance to extraction.
  • Phenolic antioxidant for thermo-oxidative stabilization of polymers
  • Improves resistance to water extraction and reduces discoloration
  • Used as processing aid and stabilizer in polyolefins and other plastics
  • Typical use levels around 0.5-2.5% in formulations
  • Solid at room temperature; molecular weight 775.2 g/mol
Catalog No. 50-002-33192