Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5,-Trimethylpyrazole, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
1072-91-9
C6H10N2
110.16
MFCD00015536
HNOQAFMOBRWDKQ-UHFFFAOYSA-N
1,3,5-trimethyl-1h-pyrazole,1h-pyrazole, 1,3,5-trimethyl,pyrazole, 1,3,5-trimethyl,akos pao-1133,pubchem20856,vitas-bb tbb000655,buttpark 15357-40,ksc490o8l,1,3,5-trimethylpyrazole,1,3,5-trimethyl-1h-pyrazole #
14081
1,3,5-trimethylpyrazole
CC1=CC(=NN1C)C
1072-91-9
C6H10N2
110.16
MFCD00015536
HNOQAFMOBRWDKQ-UHFFFAOYSA-N
1,3,5-trimethyl-1h-pyrazole,1h-pyrazole, 1,3,5-trimethyl,pyrazole, 1,3,5-trimethyl,akos pao-1133,pubchem20856,vitas-bb tbb000655,buttpark 15357-40,ksc490o8l,1,3,5-trimethylpyrazole,1,3,5-trimethyl-1h-pyrazole #
14081
1,3,5-trimethylpyrazole
CC1=CC(=NN1C)C
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color White
Melting Point 35°C to 38°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 110.16
Physical Form Crystalline Mass
Percent Purity ≥98.5%
Chemical Name or Material 1,3,5,-Trimethylpyrazole, 99%
Show More Show Less