Alfa Aesar™ 1,3,5-Tris(4-bromophenyl)benzene, 97%

Manufacturer: Alfa Aesar™ H3763606

Quantity 5g

 View more versions of this product

Catalog No. AAH3763606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7511-49-1
    Formula Weight 543.1
    MDL Number MFCD00362911
    Chemical Name or Material 1,3,5-Tris(4-bromophenyl)benzene

    Specifications

    Melting Point 261° to 265°C
    Formula Weight 543.1
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    MDL Number MFCD00362911
    Quantity 5g
    Color Yellow to pale brown
    CAS 7511-49-1
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318
    Causes serious eye damage.
    Physical Form Crystalline Powder
    Chemical Name or Material 1,3,5-Tris(4-bromophenyl)benzene
    Assay 97%

    GHS H Statement
    H318
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit