Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-acetonedicarboxylic acid, 96%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic drum
542-05-2
C5H6O5
146.1
MFCD00002711
OXTNCQMOKLOUAM-UHFFFAOYSA-N
1,3-acetonedicarboxylic acid,3-oxoglutaric acid,acetonedicarboxylic acid,pentanedioic acid, 3-oxo,acetone-1,3-dicarboxylic acid,3-ketoglutaric acid,unii-ih7p7wo21p,1,3-acetonedicarboxylic acid; 3-ketoglutaric acid,ih7p7wo21p,acetondicarbonsaure
68328
3-oxopentanedioic acid
C(C(=O)CC(=O)O)C(=O)O
542-05-2
C5H6O5
146.1
MFCD00002711
OXTNCQMOKLOUAM-UHFFFAOYSA-N
1,3-acetonedicarboxylic acid,3-oxoglutaric acid,acetonedicarboxylic acid,pentanedioic acid, 3-oxo,acetone-1,3-dicarboxylic acid,3-ketoglutaric acid,unii-ih7p7wo21p,1,3-acetonedicarboxylic acid; 3-ketoglutaric acid,ih7p7wo21p,acetondicarbonsaure
68328
3-oxopentanedioic acid
C(C(=O)CC(=O)O)C(=O)O
Flash Point >110°C
Melting Point 133°C
Infrared Spectrum Authentic
Water 2% Max. (K.F.)
Quantity 2.5kg
Packaging Plastic drum
Color White
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula HO2CCH2COCH2CO2H
Beilstein 03,789
Fieser 11,2
Merck Index 15,67
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ethyl acetate and ether,insoluble in benzene,chloroform,ligroin,decomposes in hot water,acids,alkalies to co2 an,d acetone
Formula Weight 146.1
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material 1, 3-Acetonedicarboxylic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth

GHS Signal Word: Warning

EINECSNumber : 208-797-9

Recommended Storage : preferably stored under cool conditions