Alfa Aesar™ 1,3-Benzenediboronic acid, 97%

Manufacturer: Alfa Aesar™ B2490314

Quantity 25g

 View more versions of this product

Catalog No. AAB2490314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4612-28-6
    Molecular Formula C6H8B2O4
    Formula Weight 165.75
    MDL Number MFCD03092915
    Synonym 1,3-Phenylenediboronic acid
    Chemical Name or Material 1,3-Benzenediboronic acid

    Specifications

    Formula Weight 165.75
    Molecular Formula C6H8B2O4
    Melting Point >300°C
    MDL Number MFCD03092915
    Quantity 25g
    CAS 4612-28-6
    Synonym 1,3-Phenylenediboronic acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1,3-Benzenediboronic acid
    Assay 97%

    Description

    Applications
    1,3-benzenediboronic acid is used to prepare 1,3-didydrxybenzene by oxidation reaction using alkaline hydrogen peroxide.

    Solubility
    Slightly soluble in water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit