Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Benzenediboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
4612-28-6
C6H8B2O4
165.746
MFCD03092915
UXPAASVRXBULRG-UHFFFAOYSA-N
1,3-benzenediboronic acid,1,3-phenylenediboronic acid,1,3-benzendiboronic acid,1,3-benzenediboronicacid,m-phenylenediboronic acid,3-dihydroxyboranyl phenyl boronic acid,3-dihydroxyboranyl phenylboronic acid,m-benzenediboronic acid,pubchem17415,1,3-benzendibororic acid
230477
(3-boronophenyl)boronic acid
B(C1=CC(=CC=C1)B(O)O)(O)O
4612-28-6
C6H8B2O4
165.746
MFCD03092915
UXPAASVRXBULRG-UHFFFAOYSA-N
1,3-benzenediboronic acid,1,3-phenylenediboronic acid,1,3-benzendiboronic acid,1,3-benzenediboronicacid,m-phenylenediboronic acid,3-dihydroxyboranyl phenyl boronic acid,3-dihydroxyboranyl phenylboronic acid,m-benzenediboronic acid,pubchem17415,1,3-benzendibororic acid
230477
(3-boronophenyl)boronic acid
B(C1=CC(=CC=C1)B(O)O)(O)O
Applications
1,3-benzenediboronic acid is used to prepare 1,3-didydrxybenzene by oxidation reaction using alkaline hydrogen peroxide.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
Quantity 1g
Melting Point >300°C
Formula Weight 165.75
Percent Purity 97%
Chemical Name or Material 1,3-Benzenediboronic acid

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger