Learn More
1,3-Benzenedimethanol, 98%, Thermo Scientific Chemicals
Description
1,3-Benzenedimethanol was used in the synthesis of a series of cross-linked poly(orthocarbonate)s. It was also used in the synthesis of mixed- tethered system, required for the preparation of e-edge-[60]fullerenylmethanodihydropyrrole adduct.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Benzenedimethanol was used in the synthesis of a series of cross-linked poly(orthocarbonate)s. It was also used in the synthesis of mixed- tethered system, required for the preparation of e-edge-[60]fullerenylmethanodihydropyrrole adduct.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Specifications
Melting Point | 56°C to 60°C |
Boiling Point | 154°C to 159°C (13 mmHg) |
Quantity | 5 g |
Beilstein | 1562398 |
Solubility Information | Soluble in water. |
Formula Weight | 138.17 |
Percent Purity | 98% |
Chemical Name or Material | 1,3-Benzenedimethanol |
Safety and Handling
Hazard Category | H302-H319 |
Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
EINECSNumber | 210-934-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.