Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Benzenedimethanol, 98%

Click to view available options
Quantity:
1g
5g
25g
626-18-6
C8H10O2
138.166
MFCD00004649
YWMLORGQOFONNT-UHFFFAOYSA-N
1,3-benzenedimethanol,3-hydroxymethyl phenyl methanol,1,3-phenylenedimethanol,1,3-dihydroxymethylbenzene,m-xylene-alpha,alpha'-diol,m-xylene glycol,m-xylylene glycol,3-hydroxymethyl phenyl methan-1-ol,m-xylylenalkohol,m-benzenedimethanol
69374
[3-(hydroxymethyl)phenyl]methanol
C1=CC(=CC(=C1)CO)CO
626-18-6
C8H10O2
138.166
MFCD00004649
YWMLORGQOFONNT-UHFFFAOYSA-N
1,3-benzenedimethanol,3-hydroxymethyl phenyl methanol,1,3-phenylenedimethanol,1,3-dihydroxymethylbenzene,m-xylene-alpha,alpha'-diol,m-xylene glycol,m-xylylene glycol,3-hydroxymethyl phenyl methan-1-ol,m-xylylenalkohol,m-benzenedimethanol
69374
[3-(hydroxymethyl)phenyl]methanol
C1=CC(=CC(=C1)CO)CO
Applications
1,3-Benzenedimethanol was used in the synthesis of a series of cross-linked poly(orthocarbonate)s. It was also used in the synthesis of mixed- tethered system, required for the preparation of e-edge-[60]fullerenylmethanodihydropyrrole adduct.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Melting Point 56°C to 60°C
Boiling Point 154°C to 159°C (13mmHg)
Quantity 25g
Beilstein 1562398
Solubility Information Soluble in water.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,3-Benzenedimethanol

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

EINECSNumber : 210-934-2

TSCA : Yes

Recommended Storage : Ambient temperatures