Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzenedimethanol, 98%, ACROS Organics™

Click to view available options
Quantity:
2g
10g
Packaging:
Glass bottle
626-18-6
C8H10O2
138.17
MFCD00004649
YWMLORGQOFONNT-UHFFFAOYSA-N
1,3-benzenedimethanol, 3-hydroxymethyl phenyl methanol, 1,3-phenylenedimethanol, 1,3-dihydroxymethylbenzene, m-xylene-alpha,alpha'-diol, m-xylene glycol, m-xylylene glycol, 3-hydroxymethyl phenyl methan-1-ol, m-xylylenalkohol, m-benzenedimethanol
69374
[3-(hydroxymethyl)phenyl]methanol
C1=CC(=CC(=C1)CO)CO
626-18-6
C8H10O2
138.17
MFCD00004649
YWMLORGQOFONNT-UHFFFAOYSA-N
1,3-benzenedimethanol, 3-hydroxymethyl phenyl methanol, 1,3-phenylenedimethanol, 1,3-dihydroxymethylbenzene, m-xylene-alpha,alpha'-diol, m-xylene glycol, m-xylylene glycol, 3-hydroxymethyl phenyl methan-1-ol, m-xylylenalkohol, m-benzenedimethanol
69374
[3-(hydroxymethyl)phenyl]methanol
C1=CC(=CC(=C1)CO)CO
Quantity 10g
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 56.0°C to 60.0°C
Boiling Point 154.0°C to 159.0°C (13.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H4(CH2OH)2
Beilstein 06, 914
Formula Weight 138.17
Physical Form Solution, may develop some turbidity precipitate
Percent Purity 98%
Chemical Name or Material 1, 3-Benzenedimethanol, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 210-934-2

TSCA : TSCA