Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzothiazol-6-ylmethylamine dihydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1375964-49-0
C8H8N22ClH
237.15
KFQZRIYKOSJQHF-UHFFFAOYSA-N
6-benzothiazolemethanamine dihydrochloride,1,3-benzothiazol-6-ylmethylamine dihydrochloride,1-1,3-benzothiazol-6-yl methanamine dihydrochloride
110210982
1,3-benzothiazol-6-ylmethanamine;dihydrochloride
C1=CC2=C(C=C1CN)SC=N2.Cl.Cl
1375964-49-0
C8H8N22ClH
237.15
KFQZRIYKOSJQHF-UHFFFAOYSA-N
6-benzothiazolemethanamine dihydrochloride,1,3-benzothiazol-6-ylmethylamine dihydrochloride,1-1,3-benzothiazol-6-yl methanamine dihydrochloride
110210982
1,3-benzothiazol-6-ylmethanamine;dihydrochloride
C1=CC2=C(C=C1CN)SC=N2.Cl.Cl
Quantity 5g
Melting Point 137.0°C to 137.5°C
Assay Percent Range 96 to max. 104% min. (Argentometry)
Formula Weight 237.15
Percent Purity 97%
Chemical Name or Material 1, 3-Benzothiazol-6-ylmethylamine dihydrochloride

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF

GHS Signal Word: Warning