Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzothiazole-6-carboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
19989-67-4
C8H5NOS
163.2
AVSFPLJXSHRMHM-UHFFFAOYSA-N
6-benzothiazolecarboxaldehyde,benzo d thiazole-6-carbaldehyde,1,3-benzothiazole-6-carboxaldehyde,6-formylbenzothiazole,benzothiazol-6-carboxaldehyde,6-benzothiazolecarbaldehyde,1?3-benzothiazole-6-carboxaldehyde,6-benzo d thiazolecarboxaldehyde,6-benzothiazolecarboxaldehyde 8ci,9ci
15089709
1,3-benzothiazole-6-carbaldehyde
C1=CC2=C(C=C1C=O)SC=N2
19989-67-4
C8H5NOS
163.2
AVSFPLJXSHRMHM-UHFFFAOYSA-N
6-benzothiazolecarboxaldehyde,benzo d thiazole-6-carbaldehyde,1,3-benzothiazole-6-carboxaldehyde,6-formylbenzothiazole,benzothiazol-6-carboxaldehyde,6-benzothiazolecarbaldehyde,1?3-benzothiazole-6-carboxaldehyde,6-benzo d thiazolecarboxaldehyde,6-benzothiazolecarboxaldehyde 8ci,9ci
15089709
1,3-benzothiazole-6-carbaldehyde
C1=CC2=C(C=C1C=O)SC=N2
Melting Point 92.0°C to 93.0°C
Quantity 5g
Color Brown to White
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 163.2
Physical Form Crystalline Powder or Low Melting Solid
Percent Purity 97%
Chemical Name or Material 1, 3-Benzothiazole-6-carboxaldehyde
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning