Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(1-adamantyl)benzimidazolium chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
500mg
1g
852634-41-4
C27H35N2·Cl
423.05
GMYUUHXEUIPSKA-UHFFFAOYSA-M
1,3-bis 1-adamanthyl benzimidazolium chloride,1,3-bis 1-adamantyl benzimidazolium chloride,1,3-bis 1-adamantyl-benzimidazolium chloride,1,3-di adamantan-1-yl-1h-benzo d imidazol-3-ium chloride,1,3-bis adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride,1,3-bis adamantan-1-yl-2h-1,3-benzodiazol-2-ylium chloride,1,3-bis 3r,5s,7s-adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride
16218257
1,3-bis(1-adamantyl)benzimidazol-3-ium;chloride
C1C2CC3CC1CC(C2)(C3)N4C=[N+](C5=CC=CC=C54)C67CC8CC(C6)CC(C8)C7.[Cl-]
852634-41-4
C27H35N2·Cl
423.05
GMYUUHXEUIPSKA-UHFFFAOYSA-M
1,3-bis 1-adamanthyl benzimidazolium chloride,1,3-bis 1-adamantyl benzimidazolium chloride,1,3-bis 1-adamantyl-benzimidazolium chloride,1,3-di adamantan-1-yl-1h-benzo d imidazol-3-ium chloride,1,3-bis adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride,1,3-bis adamantan-1-yl-2h-1,3-benzodiazol-2-ylium chloride,1,3-bis 3r,5s,7s-adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride
16218257
1,3-bis(1-adamantyl)benzimidazol-3-ium;chloride
C1C2CC3CC1CC(C2)(C3)N4C=[N+](C5=CC=CC=C54)C67CC8CC(C6)CC(C8)C7.[Cl-]
Melting Point 283.9°C to 285.6°C
Quantity 500mg
Color Gray to White
Infrared Spectrum Authentic
Elemental Analysis 72.06 to 81.26% C, 6.22 to 7.02% N
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 423.05
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 1, 3-Bis(1-Adamantyl)benzimidazolium chloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning