Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-bis(2,4,6-trimethylphenyl)imidazolidinium chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
173035-10-4
C21H27ClN2
342.91
MFCD09039279
1,3-dimesitylimidazolidine hydrochloride,1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride,1,3-dimesitylimidazolidine, chloride salt,1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride,1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride
173035-10-4
C21H27ClN2
342.91
MFCD09039279
1,3-dimesitylimidazolidine hydrochloride,1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride,1,3-dimesitylimidazolidine, chloride salt,1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride,1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride
Melting Point 280°C to 286°C
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color Beige to Brown-Pink
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 342.91
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 1,3-Bis(2,4,6-trimethylphenyl)imidazolidinium chloride
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning