Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
141556-45-8
C21H25ClN2
340.9
MFCD02684541
OTOSIXGMLYKKOW-UHFFFAOYSA-M
1,3-bis 2,4,6-trimethylphenyl imidazolium chloride,1,3-dimesityl-1h-imidazol-3-ium chloride,1,3-dimesitylimidazolium chloride,1,3-bis mesityl imidazolium chloride,imes.hcl,1,3-bis 2,4,6-trimethylphenyl imidazoliumchloride,1,3-2,4,6-trimethylphenyl imidazolium chloride,1,3-bis-2,4,6-trimethylphenyl-1h-imidazolium chloride,1,3-bis-2,4,6-trimethyl-phenyl-3h-imidazol-1-ium chloride,1,3-dihydro-1,3-dimesityl-2h-imidazol-2-ylidene monohydrochloride
2734211
1,3-bis(2,4,6-trimethylphenyl)imidazol-1-ium;chloride
CC1=CC(=C(C(=C1)C)N2C=C[N+](=C2)C3=C(C=C(C=C3C)C)C)C.[Cl-]
141556-45-8
C21H25ClN2
340.9
MFCD02684541
OTOSIXGMLYKKOW-UHFFFAOYSA-M
1,3-bis 2,4,6-trimethylphenyl imidazolium chloride,1,3-dimesityl-1h-imidazol-3-ium chloride,1,3-dimesitylimidazolium chloride,1,3-bis mesityl imidazolium chloride,imes.hcl,1,3-bis 2,4,6-trimethylphenyl imidazoliumchloride,1,3-2,4,6-trimethylphenyl imidazolium chloride,1,3-bis-2,4,6-trimethylphenyl-1h-imidazolium chloride,1,3-bis-2,4,6-trimethyl-phenyl-3h-imidazol-1-ium chloride,1,3-dihydro-1,3-dimesityl-2h-imidazol-2-ylidene monohydrochloride
2734211
1,3-bis(2,4,6-trimethylphenyl)imidazol-1-ium;chloride
CC1=CC(=C(C(=C1)C)N2C=C[N+](=C2)C3=C(C=C(C=C3C)C)C)C.[Cl-]
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (Argentometry)
Formula Weight 340.9
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning