Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2,6-diisopropylphenyl)-1,3,2-diazaphospholidine 2-Oxide 98.0+%, TCI America™

Click to view available options
Quantity:
200mg
1g
854929-36-5
C26H38N2OP+
425.577
PJGJPWAXZUCDHL-UHFFFAOYSA-N
12183214
1,3-bis[2,6-di(propan-2-yl)phenyl]-1,3,2-diazaphospholidin-2-ium 2-oxide
CC(C)C1=C(C(=CC=C1)C(C)C)N2CCN([P+]2=O)C3=C(C=CC=C3C(C)C)C(C)C
854929-36-5
C26H38N2OP+
425.577
PJGJPWAXZUCDHL-UHFFFAOYSA-N
12183214
1,3-bis[2,6-di(propan-2-yl)phenyl]-1,3,2-diazaphospholidin-2-ium 2-oxide
CC(C)C1=C(C(=CC=C1)C(C)C)N2CCN([P+]2=O)C3=C(C=CC=C3C(C)C)C(C)C
Color White-Yellow
Quantity 200mg
Melting Point 203°C
Formula Weight 426.58
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (N)
Chemical Name or Material 1,3-Bis(2,6-diisopropylphenyl)-1,3,2-diazaphospholidine 2-Oxide