Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
258278-25-0
C27H39ClN2
427.07
MFCD07369796
LWPXTYZKAWSRIP-UHFFFAOYSA-M
1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium chloride,sipr.hcl,1,3-bis 2,6-diisopropylphenyl-imidazolidinium-chloride,1,3-bis 2,6-diisopropylphenyl imidazolidinium chloride,n,n'-2,6-diisopropylphenyl dihydroimidazolium chloride,1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride,1,3-bis 2,6-diisopropylphenyl imidazolinium chloride?,1,3-bis-2,6-diisopropylphenyl imidazolidinium chloride,1,3-bis-2,6-diisopropylphenyl imidazolidiniumchloride,1,3-bis 2,6-bis isopropyl phenyl imidazolin-3-ium chloride
2734918
1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;chloride
CC(C)C1=C(C(=CC=C1)C(C)C)N2CC[N+](=C2)C3=C(C=CC=C3C(C)C)C(C)C.[Cl-]
258278-25-0
C27H39ClN2
427.07
MFCD07369796
LWPXTYZKAWSRIP-UHFFFAOYSA-M
1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium chloride,sipr.hcl,1,3-bis 2,6-diisopropylphenyl-imidazolidinium-chloride,1,3-bis 2,6-diisopropylphenyl imidazolidinium chloride,n,n'-2,6-diisopropylphenyl dihydroimidazolium chloride,1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride,1,3-bis 2,6-diisopropylphenyl imidazolinium chloride?,1,3-bis-2,6-diisopropylphenyl imidazolidinium chloride,1,3-bis-2,6-diisopropylphenyl imidazolidiniumchloride,1,3-bis 2,6-bis isopropyl phenyl imidazolin-3-ium chloride
2734918
1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;chloride
CC(C)C1=C(C(=CC=C1)C(C)C)N2CC[N+](=C2)C3=C(C=CC=C3C(C)C)C(C)C.[Cl-]
Infrared Spectrum Authentic
Quantity 5g
Color White to Yellow
Packaging Glass bottle
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Formula Weight 427.07
Physical Form Crystalline Powder
Percent Purity ≥88%
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium chloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.