Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium tetrafluoroborate, 97+%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
282109-83-5
C27H39BF4N2
478.42
MFCD04971187
KFZBJQUHHALFSR-UHFFFAOYSA-N
1,3-bis 2,6-diisopropylphenyl-4,5-dihydroimidazolium tetrafluoroborate,1,3-bis 2,6-di-i-propylphenyl-4,5-dihydroimidazolium tetrafluoroborate,sipr-hbf4,1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium tetrafluoroborate,4,5-dihydro-1,3-bis 2,6-diisopropylphenyl imidazolium tetrafluoroborate,n,n'-bis 2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate,sipr.hbf4,1,3-bis-2,6-diisopropylphenyl imidazolinium tetrafluoroborate,n,n'-2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate,1,3-bis 2,6-di-i-propylphenyl-4,5-dihydro-imidazolium tetra-fluoroborate
11375006
1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)C1=C(C(=CC=C1)C(C)C)N2CC[N+](=C2)C3=C(C=CC=C3C(C)C)C(C)C
282109-83-5
C27H39BF4N2
478.42
MFCD04971187
KFZBJQUHHALFSR-UHFFFAOYSA-N
1,3-bis 2,6-diisopropylphenyl-4,5-dihydroimidazolium tetrafluoroborate,1,3-bis 2,6-di-i-propylphenyl-4,5-dihydroimidazolium tetrafluoroborate,sipr-hbf4,1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium tetrafluoroborate,4,5-dihydro-1,3-bis 2,6-diisopropylphenyl imidazolium tetrafluoroborate,n,n'-bis 2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate,sipr.hbf4,1,3-bis-2,6-diisopropylphenyl imidazolinium tetrafluoroborate,n,n'-2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate,1,3-bis 2,6-di-i-propylphenyl-4,5-dihydro-imidazolium tetra-fluoroborate
11375006
1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)C1=C(C(=CC=C1)C(C)C)N2CC[N+](=C2)C3=C(C=CC=C3C(C)C)C(C)C
Elemental Analysis 97.0% min.
Infrared Spectrum Authentic
Quantity 5g
Color White
Packaging Glass bottle
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Formula Weight 478.42
Physical Form Powder
Percent Purity 97+%
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium tetrafluoroborate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning