Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate, 98%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
915967-82-7
C29H52Cl2N6O20P4
999.552
MFCD11870901
AMCQDGFOKTXHSY-UHFFFAOYSA-N
piperaquine phosphate hydrate,piperaquine tetraphosphate tetrahydrate,unii-854d7k8lxb,piperaquine tetraphosphate hydrate,piperaquine phosphate,quinoline, 4,4'-1,3-propanediyldi-4,1-piperazinediyl bis 7-chloro-, phosphate, hydrate 1:4:4,piperaquine tetraphosphate tetrahydrate hplc,tetrakis phosphoric acid piperaquine tetrahydrate,1,3-bis 4-7-chloroquinolin-4-yl piperazin-1-yl propane tetrakis phosphate tetrahydrate
49849842
7-chloro-4-[4-[3-[4-(7-chloroquinolin-4-yl)piperazin-1-yl]propyl]piperazin-1-yl]quinoline;phosphoric acid;tetrahydrate
C1CN(CCN1CCCN2CCN(CC2)C3=C4C=CC(=CC4=NC=C3)Cl)C5=C6C=CC(=CC6=NC=C5)Cl.O.O.O.O.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O
915967-82-7
C29H52Cl2N6O20P4
999.552
MFCD11870901
AMCQDGFOKTXHSY-UHFFFAOYSA-N
piperaquine phosphate hydrate,piperaquine tetraphosphate tetrahydrate,unii-854d7k8lxb,piperaquine tetraphosphate hydrate,piperaquine phosphate,quinoline, 4,4'-1,3-propanediyldi-4,1-piperazinediyl bis 7-chloro-, phosphate, hydrate 1:4:4,piperaquine tetraphosphate tetrahydrate hplc,tetrakis phosphoric acid piperaquine tetrahydrate,1,3-bis 4-7-chloroquinolin-4-yl piperazin-1-yl propane tetrakis phosphate tetrahydrate
49849842
7-chloro-4-[4-[3-[4-(7-chloroquinolin-4-yl)piperazin-1-yl]propyl]piperazin-1-yl]quinoline;phosphoric acid;tetrahydrate
C1CN(CCN1CCCN2CCN(CC2)C3=C4C=CC(=CC4=NC=C3)Cl)C5=C6C=CC(=CC6=NC=C5)Cl.O.O.O.O.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O
Sensitivity Hygroscopic
Quantity 100g
Color White
Formula Weight 999.55
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning