Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-bis(dicyclohexylphosphino)propane, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
103099-52-1
C27H50P2
436.64
MFCD00799482
RSJBEKXKEUQLER-UHFFFAOYSA-N
1,3-bis dicyclohexylphosphino propane,phosphine, 1,3-propanediylbis dicyclohexyl,dicyclohexyl 3-dicyclohexylphosphanylpropyl phosphane,1,3-bis dicyclohexylphosphonium propane bis tetrafluoroborate,acmc-1butf,dicyclohexyl 3-dicyclohexylphosphino propyl phosphine #
438876
dicyclohexyl(3-dicyclohexylphosphanylpropyl)phosphane
C1CCC(CC1)P(CCCP(C2CCCCC2)C3CCCCC3)C4CCCCC4
103099-52-1
C27H50P2
436.64
MFCD00799482
RSJBEKXKEUQLER-UHFFFAOYSA-N
1,3-bis dicyclohexylphosphino propane,phosphine, 1,3-propanediylbis dicyclohexyl,dicyclohexyl 3-dicyclohexylphosphanylpropyl phosphane,1,3-bis dicyclohexylphosphonium propane bis tetrafluoroborate,acmc-1butf,dicyclohexyl 3-dicyclohexylphosphino propyl phosphine #
438876
dicyclohexyl(3-dicyclohexylphosphanylpropyl)phosphane
C1CCC(CC1)P(CCCP(C2CCCCC2)C3CCCCC3)C4CCCCC4
Color Colorless
Quantity 1g
Boiling Point 350.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point >110°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Solubility Information Solubility in water: insoluble
Formula Weight 436.64
Physical Form Paste
Percent Purity 95%
Chemical Name or Material 1, 3-Bis(dicyclohexylphosphino)propane
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning