Alfa Aesar™ 1,3-Bis(methylthio)-2-propanol, 97%

Manufacturer: Alfa Aesar™ B2189306

Quantity 5g

 View more versions of this product

Catalog No. AAB2189306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 31805-83-1
    Molecular Formula CH3SCH2CH(OH)CH2SCH3
    Formula Weight 152.29
    MDL Number MFCD00041435
    Beilstein 1734314
    Chemical Name or Material 1,3-Bis(methylthio)-2-propanol

    Specifications

    Formula Weight 152.29
    Molecular Formula CH3SCH2CH(OH)CH2SCH3
    MDL Number MFCD00041435
    CAS 31805-83-1
    Beilstein 1734314
    Boiling Point 112° to 114°C (5mmHg)
    Quantity 5g
    Chemical Name or Material 1,3-Bis(methylthio)-2-propanol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit