Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Bis(methylthio)-2-propanol, 97%

Click to view available options
Quantity:
1g
5g
31805-83-1
C5H12OS2
152.27
MFCD00041435
BTQNSLFPJKRBQK-UHFFFAOYSA-N
1,3-bis methylthio-2-propanol,1,3-bis methylthio propan-2-ol,1,3-bis methylsulfanyl propan-2-ol,1,3-bis methylsulfanyl-2-propanol,2-propanol, 1,3-bis methylthio,acmc-20ap53,2-propanol,1,3-bis methylthio,1,3-bis methylsulfanyl-propan-2-ol,1,3-bis methylsulfanyl-2-propanol #
99550
1,3-bis(methylsulfanyl)propan-2-ol
CSCC(CSC)O
31805-83-1
C5H12OS2
152.27
MFCD00041435
BTQNSLFPJKRBQK-UHFFFAOYSA-N
1,3-bis methylthio-2-propanol,1,3-bis methylthio propan-2-ol,1,3-bis methylsulfanyl propan-2-ol,1,3-bis methylsulfanyl-2-propanol,2-propanol, 1,3-bis methylthio,acmc-20ap53,2-propanol,1,3-bis methylthio,1,3-bis methylsulfanyl-propan-2-ol,1,3-bis methylsulfanyl-2-propanol #
99550
1,3-bis(methylsulfanyl)propan-2-ol
CSCC(CSC)O
Boiling Point 112°C to 114°C (5mmHg)
Quantity 1g
Beilstein 1734314
Formula Weight 152.29
Percent Purity 97%
Chemical Name or Material 1,3-Bis(methylthio)-2-propanol

EINECSNumber : 250-814-7

TSCA : No

Recommended Storage : Ambient temperatures