Alfa Aesar™ 1,3-Bis(methylthio)propane, 97%

Manufacturer: Alfa Aesar™ B2239703

Quantity 1g

 View more versions of this product

Catalog No. AAB2239703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24949-35-7
    Molecular Formula CH3S(CH2)3SCH3
    Formula Weight 136.28
    MDL Number MFCD00039811
    Beilstein 1732333
    Synonym 1,3-Bis(methylthio)propane
    Chemical Name or Material 1,3-Bis(methylthio)propane

    Specifications

    Formula Weight 136.28
    Molecular Formula CH3S(CH2)3SCH3
    MDL Number MFCD00039811
    CAS 24949-35-7
    Beilstein 1732333
    Synonym 1,3-Bis(methylthio)propane
    Boiling Point 88° to 90°C (12mmHg)
    Quantity 1g
    Flash Point >100°C (212°F)
    Odor Stench
    Chemical Name or Material 1,3-Bis(methylthio)propane
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit