Alfa Aesar™ 1,3-Bis(trifluoromethyl)benzene, 98+%

Manufacturer: Alfa Aesar™ A1136030

Quantity 250g

 View more versions of this product

Catalog No. AAA1136030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 402-31-3
    Molecular Formula C8H4F6
    Formula Weight 214.11
    MDL Number MFCD00000392
    UN Number UN1993
    Beilstein 2052589
    Synonym alpha,alpha,alpha,alpha',alpha',alpha'-Hexafluoro-m-xylene
    Chemical Name or Material 1,3-Bis (trifluoromethyl)benzene

    Specifications

    Formula Weight 214.11
    Health Hazard 3 GHS P Statement
    P210-P260-P280g-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Boiling Point 115° to 117°C
    Health Hazard 1 Warning
    TSCA TSCA
    Molecular Formula C8H4F6
    CAS 402-31-3
    Odor Aromatic
    Beilstein 2052589
    Synonym alpha,alpha,alpha,alpha',alpha',alpha'-Hexafluoro-m-xylene
    Melting Point −35°C
    Quantity 250g
    Flash Point 26°C (78°F)
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    MDL Number MFCD00000392
    UN Number UN1993
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 1,3-Bis (trifluoromethyl)benzene
    Assay ≥98%

    Description

    Applications
    1,3-Bis(trifluoromethyl)benzene undergoes regioselective metalation and subsequent carboxylation at position 2 to give 2,6-bis(trifluoromethyl)benzoic acid. a convenient, selective synthesis of bis[2,4-bis(trifluoromethyl)phenyl]phosphane derivatives with 1,3-bis(trifluoromethyl)benzene as starting material.

    Solubility
    Insoluble in water. Soluble in alcohol, ether, benzene.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P280g-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit