Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis[tris(hydroxymethyl)amino]propane, 99%, Acros Organics™

Click to view available options
Quantity:
25g
100g
250g
1kg
Packaging:
Glass Bottle
64431-96-5
C11H26N2O6
282.33
MFCD00004689
HHKZCCWKTZRCCL-UHFFFAOYSA-N
bis-tris propane,1,3-bis tris hydroxymethyl methylamino propane,2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol,2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol,1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl,2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol,2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol,b3p
125132
CHEBI:40947
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
64431-96-5
C11H26N2O6
282.33
MFCD00004689
HHKZCCWKTZRCCL-UHFFFAOYSA-N
bis-tris propane,1,3-bis tris hydroxymethyl methylamino propane,2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol,2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol,1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl,2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol,2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol,b3p
125132
CHEBI:40947
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
Packaging Glass Bottle
Elemental Analysis 46.33 to 47.2% C, 9.82 to 10.02% N
Melting Point 164.0°C to 165.0°C
Infrared Spectrum Authentic
Color White
Quantity 1kg
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH2{CH2NHC(CH2OH)3]2
Beilstein 04,III,859
Solubility Information Solubility in water: soluble in cold water
Formula Weight 282.33
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 1,3-Bis[tris(hydroxymethyl)amino]propane
Show More Show Less

EINECSNumber : 264-899-3