Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Bromopropoxy)-2-bromobenzene, 93%, ACROS Organics™

Click to view available options
Quantity:
25g
37136-84-8
C9H10Br2O
293.986
MFCD02030868
XREHAMWVAOBJBV-UHFFFAOYSA-N
1-bromo-2-3-bromopropoxy benzene,1-3-bromopropoxy-2-bromobenzene,3-2-bromophenoxy bromopropane,2-bromophenyl 3-bromopropyl ether,benzene,1-bromo-2-3-bromopropoxy
334092
1-bromo-2-(3-bromopropoxy)benzene
C1=CC=C(C(=C1)OCCCBr)Br
37136-84-8
C9H10Br2O
293.986
MFCD02030868
XREHAMWVAOBJBV-UHFFFAOYSA-N
1-bromo-2-3-bromopropoxy benzene,1-3-bromopropoxy-2-bromobenzene,3-2-bromophenoxy bromopropane,2-bromophenyl 3-bromopropyl ether,benzene,1-bromo-2-3-bromopropoxy
334092
1-bromo-2-(3-bromopropoxy)benzene
C1=CC=C(C(=C1)OCCCBr)Br
Color Yellow
Quantity 25g
Infrared Spectrum Authentic
Refractive Index 1.5760 to 1.5780 (20°C, 589nm)
CAS Min % 91.0
CAS Max % 100.0
Assay Percent Range 93%
Linear Formula BrC6H4O(CH2)3Br
Formula Weight 293.99
Physical Form Liquid
Percent Purity ≥91%
Chemical Name or Material 1-(3-Bromopropoxy)-2-bromobenzene
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.