Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-bromopropoxy)-2-nitrobenzene, 90+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
104147-69-5
C9H10BrNO3
260.09
MFCD00596660
HPZBIRIHQKSGGT-UHFFFAOYSA-N
1-3-bromopropoxy-2-nitrobenzene,1-3-bromo-propoxy-2-nitro-benzene
7172300
1-(3-bromopropoxy)-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])OCCCBr
104147-69-5
C9H10BrNO3
260.09
MFCD00596660
HPZBIRIHQKSGGT-UHFFFAOYSA-N
1-3-bromopropoxy-2-nitrobenzene,1-3-bromo-propoxy-2-nitro-benzene
7172300
1-(3-bromopropoxy)-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])OCCCBr
Packaging Glass bottle
Quantity 5g
Flash Point 150°C
Boiling Point 131°C to 132°C (0.5mmHg)
Melting Point 36°C to 39°C
Infrared Spectrum Authentic
Color Yellow
CAS Min % 90.0
CAS Max % 100.0
Assay Percent Range 90% min. (GC)
Linear Formula NO2C6H4O(CH2)3Br
Formula Weight 260.09
Physical Form Crystalline Powder
Percent Purity 90+%
Chemical Name or Material 1-(3-Bromopropoxy)-2-nitrobenzene
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning