Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Bromopropoxy)-3-chlorobenzene, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
37142-46-4
C9H10BrClO
249.53
MFCD00596672
MVKLHPFBHSJVLA-UHFFFAOYSA-N
1-3-bromopropoxy-3-chlorobenzene,3-bromopropyl 3-chlorophenyl ether
2734929
1-(3-bromopropoxy)-3-chlorobenzene
C1=CC(=CC(=C1)Cl)OCCCBr
37142-46-4
C9H10BrClO
249.53
MFCD00596672
MVKLHPFBHSJVLA-UHFFFAOYSA-N
1-3-bromopropoxy-3-chlorobenzene,3-bromopropyl 3-chlorophenyl ether
2734929
1-(3-bromopropoxy)-3-chlorobenzene
C1=CC(=CC(=C1)Cl)OCCCBr
Refractive Index 1.5570 to 1.559
Boiling Point 139°C to 148°C (8.0mmHg)
Quantity 5g
Color Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula ClC6H4O(CH2)3Br
Formula Weight 249.53
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 1-(3-Bromopropoxy)-3-chlorobenzene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning