Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(±)-1,3-Butanediol, ≥99.0% (GC), MilliporeSigma™ Supelco™
(+/-)-1,3-Butanediol is an organic alcohol with antimicrobial activity against a wide variety of pathogens.
$85.00 - $352.00
Specifications
CAS | 107-88-0 |
---|---|
Type | Analytical Standard |
Linear Formula | CH3 CH(OH)CH2 CH2 OH |
MDL Number | MFCD00004554 |
Synonym | 1,3-Butylene glycol |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-2418
|
MilliporeSigma Supelco
090681ML |
1 mL |
Each for $85.00
|
|
|||||
11-101-2419
|
MilliporeSigma Supelco
090685ML |
5 mL |
Each for $352.00
|
|
|||||
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
107-88-0 | |
CH3 CH(OH)CH2 CH2 OH | |
1,3-Butylene glycol | |
≥99.0% (GC) | |
741°F | |
3.1 (20°C, vs. air) | |
203°C to 204°C |
Analytical Standard | |
MFCD00004554 | |
90.12 | |
Analytical Standard | |
0.06 mmHg (20°C) | |
1.005 g/mL (at 25°C) | |
108°C |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : EK0440000
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title