Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-chlorophenyl)-1-cyclopentanecarbonitrile, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
143328-16-9
C12H12ClN
205.685
MFCD08459363
VAJLOPYWHOVXCC-UHFFFAOYSA-N
1-3-chlorophenyl cyclopentanecarbonitrile,1-3-chlorophenyl cyclopentane-1-carbonitrile,1-3-chlorophenyl-1-cyclopentanecarbonitrile,cyclopentanecarbonitrile,1-3-chlorophenyl,acmc-1bwlf
15742719
1-(3-chlorophenyl)cyclopentane-1-carbonitrile
C1CCC(C1)(C#N)C2=CC(=CC=C2)Cl
143328-16-9
C12H12ClN
205.685
MFCD08459363
VAJLOPYWHOVXCC-UHFFFAOYSA-N
1-3-chlorophenyl cyclopentanecarbonitrile,1-3-chlorophenyl cyclopentane-1-carbonitrile,1-3-chlorophenyl-1-cyclopentanecarbonitrile,cyclopentanecarbonitrile,1-3-chlorophenyl,acmc-1bwlf
15742719
1-(3-chlorophenyl)cyclopentane-1-carbonitrile
C1CCC(C1)(C#N)C2=CC(=CC=C2)Cl
Color Brown
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Formula Weight 205.69
Physical Form Low Melting Mass
Percent Purity ≥98.5%
Chemical Name or Material 1-(3-Chlorophenyl)-1-cyclopentanecarbonitrile

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning