Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Cyclohexadiene, 96%, stabilized, Thermo Scientific Chemicals

Catalog Number p-96163
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
592-57-4
C6H8
80.13
MFCD00001532
MGNZXYYWBUKAII-UHFFFAOYSA-N
1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3
11605
CHEBI:37610
cyclohexa-1,3-diene
C1CC=CC=C1
592-57-4
C6H8
80.13
MFCD00001532
MGNZXYYWBUKAII-UHFFFAOYSA-N
1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3
11605
CHEBI:37610
cyclohexa-1,3-diene
C1CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point -98.0°C
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 80.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4730 to 1.4760
Quantity 5 mL
Specific Gravity 0.86
Solubility Information Solubility in water: immiscible.
Stabilizer 0.1% BHT
Formula Weight 80.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1,3-Cyclohexadiene, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
EINECSNumber 209-764-1
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit