Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Cyclopentanediol, 95%, mixture of cis and trans, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
59719-74-3
C5H10O2
102.13
MFCD00001367
NUUPJBRGQCEZSI-UHFFFAOYSA-N
1,3-cyclopentanediol,1,3-cyclopentanediol, cis,1,3-cyclopentanediol, mixture of cis and trans,1,3-cyclopentandiol,1,4-cyclopentanediol,cyclopentan-1,3-diol,3-hydroxy cylcopentanol,cyclopentane 1,3-diol,acmc-20ap4n,1,3-dihydroxycyclopentane
100165
cyclopentane-1,3-diol
C1CC(CC1O)O
59719-74-3
C5H10O2
102.13
MFCD00001367
NUUPJBRGQCEZSI-UHFFFAOYSA-N
1,3-cyclopentanediol,1,3-cyclopentanediol, cis,1,3-cyclopentanediol, mixture of cis and trans,1,3-cyclopentandiol,1,4-cyclopentanediol,cyclopentan-1,3-diol,3-hydroxy cylcopentanol,cyclopentane 1,3-diol,acmc-20ap4n,1,3-dihydroxycyclopentane
100165
cyclopentane-1,3-diol
C1CC(CC1O)O
Specific Gravity 1.09
Refractive Index 1.483
Color Colorless to Yellow
Packaging Glass bottle
Quantity 5g
Density 1.0900g/mL
Boiling Point 80.0°C to 85.0°C (0.1mmHg)
Flash Point >110°C
Melting Point 40.0°C
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula C5H8(OH)2
Beilstein 06, III, 4057
Solubility Information Solubility in water: partially soluble.
Formula Weight 102.13
Physical Form Liquid After Melting
Percent Purity 95%
Chemical Name or Material 1,3-Cyclopentanediol, Mixture of cis and trans
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 261-880-1