Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Cyclopentanedione, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3859-41-4
C5H6O2
98.1
MFCD00001405
LOGSONSNCYTHPS-UHFFFAOYSA-N
1,3-cyclopentanedione,1,3-cyclopentadione,1.3-cyclopentanedione,cyclopentan-1,3-dion,1,3cyclopentadione,1,3 cyclopentadione,1,3-cyclopentandione,1,3-dioxocyclopentane,cyclopentan-1,3-dione,1,3-cyclopentane dione
77466
CHEBI:41456
cyclopentane-1,3-dione
C1CC(=O)CC1=O
3859-41-4
C5H6O2
98.1
MFCD00001405
LOGSONSNCYTHPS-UHFFFAOYSA-N
1,3-cyclopentanedione,1,3-cyclopentadione,1.3-cyclopentanedione,cyclopentan-1,3-dion,1,3cyclopentadione,1,3 cyclopentadione,1,3-cyclopentandione,1,3-dioxocyclopentane,cyclopentan-1,3-dione,1,3-cyclopentane dione
77466
CHEBI:41456
cyclopentane-1,3-dione
C1CC(=O)CC1=O
Color Ochre to Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 148°C to 153°C
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C5H6(=O)2
Formula Weight 98.1
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 1, 3-Cyclopentanedione
Show More Show Less

EINECSNumber : 223-372-8