Alfa Aesar™ 1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane, 94%

Manufacturer: Alfa Aesar™ H2913006

Quantity 5g

 View more versions of this product

Catalog No. AAH2913006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18055-70-4
    Molecular Formula C18H26OSi2
    Formula Weight 314.57
    MDL Number MFCD09266163
    Synonym 1,3-Bis(4-methylphenyl)-1,1,3,3-tetramethyldisiloxane
    Chemical Name or Material 1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane

    Specifications

    Formula Weight 314.57
    Molecular Formula C18H26OSi2
    MDL Number MFCD09266163
    CAS 18055-70-4
    Synonym 1,3-Bis(4-methylphenyl)-1,1,3,3-tetramethyldisiloxane
    Quantity 5g
    Chemical Name or Material 1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane
    Assay 94%

    Description

    Applications
    It is employed as a intermediates for pharmaceutical.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit