Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Diacetylbenzene, 97%

Click to view available options
Quantity:
1g
5g
6781-42-6
C10H10O2
162.188
MFCD00008740
VCHOFVSNWYPAEF-UHFFFAOYSA-N
1,3-diacetylbenzene,1,1'-1,3-phenylene diethanone,ethanone, 1,1'-1,3-phenylene bis,m-diacetylbenzene,benzene-1,3-bis acetyl,m-acetylacetophenone,m-acetyl acetophenone,1-3-acetylphenyl ethanone,1-3-acetylphenyl ethan-1-one,m-diacetyl benzene
23229
1-(3-acetylphenyl)ethanone
CC(=O)C1=CC(=CC=C1)C(=O)C
6781-42-6
C10H10O2
162.188
MFCD00008740
VCHOFVSNWYPAEF-UHFFFAOYSA-N
1,3-diacetylbenzene,1,1'-1,3-phenylene diethanone,ethanone, 1,1'-1,3-phenylene bis,m-diacetylbenzene,benzene-1,3-bis acetyl,m-acetylacetophenone,m-acetyl acetophenone,1-3-acetylphenyl ethanone,1-3-acetylphenyl ethan-1-one,m-diacetyl benzene
23229
1-(3-acetylphenyl)ethanone
CC(=O)C1=CC(=CC=C1)C(=O)C
Applications
1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.

Solubility
Soluble in ethanol, benzene, chloroform.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Flash Point >110°C (230°F)
Boiling Point 150°C to 155°C (15mmHg)
Melting Point 35°C to 37°C
Quantity 5g
Beilstein 2042511
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 1,3-Diacetylbenzene