Alfa Aesar™ 1,3-Dibenzoyloxybenzene, 98%

Manufacturer: Alfa Aesar™ A1198314

Quantity 25g

 View more versions of this product

Catalog No. AAA1198314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 94-01-9
    Molecular Formula C20H14O4
    Formula Weight 318.33
    MDL Number MFCD00016576
    Beilstein 2059467
    Synonym Resorcinol dibenzoate
    Chemical Name or Material 1,3-Dibenzoyloxybenzene

    Specifications

    Formula Weight 318.33
    Quantity 25g
    Flash Point 93°C (199°F)
    CAS 94-01-9
    Beilstein 2059467
    Synonym Resorcinol dibenzoate
    Melting Point 116° to 119°C
    Molecular Formula C20H14O4
    MDL Number MFCD00016576
    RTECS Number VH0590000
    TSCA TSCA
    Chemical Name or Material 1,3-Dibenzoyloxybenzene
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit