Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dibenzoyloxybenzene, 98%

Manufacturer:  Alfa Aesar™ A1198322

Quantity 100g

 View more versions of this product

Catalog No. AAA1198322


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

94-01-9
318.33
2059467
1,3-Dibenzoyloxybenzene
C20H14O4
MFCD00016576
Resorcinol dibenzoate

Specifications

94-01-9
100g
C20H14O4
MFCD00016576
Resorcinol dibenzoate
98%
116° to 119°C
93°C (199°F)
318.33
2059467
1,3-Dibenzoyloxybenzene

Safety and Handling

SDS & Safety Documents

RTECSNumber : VH0590000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit