Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dibromo-5,5-dimethylhydantoin, 98%

Click to view available options
Quantity:
100g
500g
2500g
77-48-5
C5H6Br2N2O2
285.923
MFCD00003189
VRLDVERQJMEPIF-UHFFFAOYSA-N
1,3-dibromo-5,5-dimethylhydantoin,dibromantin,dibromantine,take charge orange,dbdmh,n,n'-dibromodimethylhydantoin,5,5-dimethyl-1,3-dibromohydantoin,2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl,1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione,unii-v9r5f9i7mz
6479
1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
CC1(C(=O)N(C(=O)N1Br)Br)C
77-48-5
C5H6Br2N2O2
285.923
MFCD00003189
VRLDVERQJMEPIF-UHFFFAOYSA-N
1,3-dibromo-5,5-dimethylhydantoin,dibromantin,dibromantine,take charge orange,dbdmh,n,n'-dibromodimethylhydantoin,5,5-dimethyl-1,3-dibromohydantoin,2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl,1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione,unii-v9r5f9i7mz
6479
1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
CC1(C(=O)N(C(=O)N1Br)Br)C
Applications
It is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.

Solubility
Soluble in Alcohol, Ether, Benzene. Insoluble in Water.

Notes
Moisture sensitive. Store away from oxidizing agents, reducing agents, flammable substances, organic materials, metal powders & moisture.
Sensitivity Moisture sensitive
Melting Point ~198°C (decomposition)
Quantity 500g
Flash Point 155°C (311°F)
UN Number UN3085
Beilstein 146024
Solubility Information Soluble in Alcohol,Ether,Benzene. Insoluble in Water.
Formula Weight 285.93
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-5,5-dimethylhydantoin

GHS H Statement
H272-H314-H318-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P210-P220-P221-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H272-H302-H314

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.

EINECSNumber : 201-030-9

RTECSNumber : MU0686000

TSCA : Yes

Recommended Storage : Ambient temperatures