Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dibromo-5-chlorobenzene, 98%

Click to view available options
Quantity:
1g
5g
25g
14862-52-3
C6H3Br2Cl
270.348
MFCD00070765
FNKCOUREFBNNHG-UHFFFAOYSA-N
3,5-dibromochlorobenzene,1-chloro-3,5-dibromobenzene,benzene, 1,3-dibromo-5-chloro,pubchem3634,ambsclk-224,intermediates-zcf02139,ksc181m3d,1,3-dibromo-5-chloro-benzene,tpc-i027,fnkcourefbnnhg-uhfffaoysa
84676
1,3-dibromo-5-chlorobenzene
C1=C(C=C(C=C1Br)Br)Cl
14862-52-3
C6H3Br2Cl
270.348
MFCD00070765
FNKCOUREFBNNHG-UHFFFAOYSA-N
3,5-dibromochlorobenzene,1-chloro-3,5-dibromobenzene,benzene, 1,3-dibromo-5-chloro,pubchem3634,ambsclk-224,intermediates-zcf02139,ksc181m3d,1,3-dibromo-5-chloro-benzene,tpc-i027,fnkcourefbnnhg-uhfffaoysa
84676
1,3-dibromo-5-chlorobenzene
C1=C(C=C(C=C1Br)Br)Cl
Melting Point 95°C to 98°C
Quantity 1g
Formula Weight 270.35
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-5-chlorobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning