Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dibromoacetone, 98%

Manufacturer:  Alfa Aesar™ H3732503

Quantity 1g

 View more versions of this product

Catalog No. AAH3732503


  Check Availability
Add to cart

Specifications

Description

Applications
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.

Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
Description & Specifications

Chemical Identifiers

816-39-7
MFCD00013540
1740389
1,3-Dibromoacetone
215.87
UN1759
1,3-Dibromo-2-propanone

Specifications

816-39-7
97° to 98°C (21mmHg)
White to pale yellow
Moisture Sensitive
MFCD00013540
1740389
1,3-Dibromoacetone
29° to 30°C
Crystals or Fused Solid
1g
215.87
UN1759
1,3-Dibromo-2-propanone
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

TSCA : TSCA

storageNote1 : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit